missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin 8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550687
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total IPO8 protein.
Importin 8 Polyclonal antibody specifically detects Importin 8 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
Importin 8 | |
Polyclonal | |
Unconjugated | |
IPO8 | |
6230418K12Rik; Abcc10; ATP-binding cassette, sub-family C (CFTR/MRP), member 10; C130009K11Rik; FLJ26580; imp8; importin 8; importin-8; IPO8; MRP7; Om1; OM-1; oocyte maturation 1; RAN binding protein 8; ran-binding protein 8; RANBP8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10526 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O15397 | |
IPO8 | |
Synthetic peptide corresponding to a region derived from internal residues of human importin 8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |