Learn More
Iprodion, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Iprodione |
| CAS | 36734-19-7 |
| Summenformel | C13 H13 Cl2 N3 O3 |
| Menge | 10 g |
| Synonym | 1-Imidazolidinecarboxamide, 3-(3,5-dichlorophenyl)-N-(1-methylethyl)-2,4-dioxo-, 3-(3,5-Dichlorophenyl)-N-(1-methylethyl)-2,4-dioxo-1-imidazolidinecarboxamide, Chipco 26019, Civet, Glycophen, Glycophene, Iprodial, Iprodin, Iprodione, Kidan, LFA 2043, Promidione, Quintalic, RP 26019, Rovral, Rovral 500 Aqua, Rovral 50WP, Rovral Aquaflo, Rovral Flo, Rovral Flo 255SC, Rovral Green GT, Rovral PM, Verisan, Xiuan |
| InChI-Formel | InChI=1S/C13H13Cl2N3O3/c1-7(2)16-12(20)17-6-11(19)18(13(17)21)10-4-8(14)3-9(15)5-10/h3-5,7H,6H2,1-2H3,(H,16,20) |
| SMILES | CC(C)NC(=O)N1CC(=O)N(C1=O)c2cc(Cl)cc(Cl)c2 |
| IUPAC-Name | 3-(3,5-dichlorphenyl)-2,4-dioxo-N-propan-2-Ylimidazolidin-1-Carbodamid |
| Molekulargewicht (g/mol) | 330.17 |
| Formelmasse | 329.0334 |