Learn More
IQSEC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558268
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GSLKRQSAYE RSLGGQQGSP KHGPHSGAPK SLPREEPELR PRPPRPLDSH LAINGSANRQ SKSESDYSDG DNDSINSTSN S Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
IQSEC1 Polyclonal antibody specifically detects IQSEC1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
IQSEC1 | |
Polyclonal | |
Unconjugated | |
IQSEC1 | |
ADP-ribosylation factors guanine nucleotide-exchange protein 100; ADP-ribosylation factors guanine nucleotide-exchange protein 2; ARFGEP100; ARF-GEP100; AW561907; BC026481; BRAG2; brefeldin A-resistant ARF-GEF2; brefeldin-resistant Arf-GEF 2 protein; cI-43; D6Ertd349e; GEP100; IQ motif and Sec7 domain 1; IQ motif and SEC7 domain-containing protein 1; Iqsec1; Kiaa0763 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9922 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6DN90 | |
IQSEC1 | |
Recombinant protein corresponding to Human IQSEC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only