missing translation for 'onlineSavingsMsg'
Learn More

IRF8 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 24437-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16882915

  • 140.00 € / 20 Mikroliter
Verfügbar ab: 04-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody specifically recognizes the 48 kDa IRF8 protein.

IRF8 (interferon regulatory factor 8) is a transcriptional activator or repressor. It specifically binds to the upstream regulatory region of type I IFN and IFN-inducible MHC class I genes. IRF8 plays a negative regulatory role in cells of the immune system. It is involved in CD8+ dendritic cell differentiation by forming a complex with the BATF-JUNB heterodimer in immune cells. This leads to recognition of AICE sequence, an immune-specific regulatory element, followed by cooperative binding of BATF and IRF8 and activation of the genes. Mutations in the gene can result in immunodeficiencey 32A and 32B.
Spezifikation

Spezifikation

IRF8
Polyclonal
Unconjugated
IRF8
H ICSBP, ICSBP, ICSBP1, interferon regulatory factor 8, IRF 8, IRF8
Rabbit
Antigen Affinity Chromatography
RUO
3394
-20°C
Liquid
Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q02556
IRF8
IRF8 Fusion Protein Ag19909
20 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote