missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisen(III)-sulfat-Hydrat, Reagenzien-Gütegrad, Thermo Scientific Chemicals
Click to view available options
Menge:
250 g
1 kg
5 kg
Beschreibung
Iron(III) sulfate is also used in pigments, pickling baths for aluminum and steel. It acts as a mordant in dyeing, as a coagulant for industrial wastes. In the medical field, it is used as an astringent and styptic. It is involved in the preparation of aromatic esters from the corresponding acids and alcohols. Further, it is employed in the oxidation of pyrite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Chemischer Name oder Material | Iron(III) sulfate hydrate |
Farbe | Grau-Weiß |
Physikalische Form | Granulatpulver |
Menge | 250 g |
Löslichkeitsinformationen | Soluble in waterSoluble in water. Sparingly soluble in alcohol. |
IUPAC-Name | Eisen(3+);trisulfat;hydrat |
Formelmasse | 399.87 (Anhydrous) |
Güte | Reagenz |
Geruch | Odorless |
Empfindlichkeit | Hygroscopic |
Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Iron(III) sulfate hydrate
- Achtung
- Hautreizende/-ätzende Wirkung Kategorie 2
- Schwere Augenschädigung/Augenreizung Kategorie 2
- Spezifische Zielorgan-Toxizität Kategorie 3
- H315-Verursacht Hautreizungen.
- H319-Verursacht schwere Augenreizung.
- H335-Kann die Atemwege reizen.
- P261-Einatmen von Staub/Rauch/Gas/Nebel/Dampf/Aerosol vermei den.
- P280-Schutzhandschuhe/Schutzkleidung/Augenschutz/Gesichtsschutz tragen.
- P305+P351+P338-BEI KONTAKT MIT DEN AUGEN: Einige Minuten lang behutsam mit Wasser spülen. Vorhandene Kontaktlinsen nach Möglichkeit entfernen. Weiter spülen.
- P304+P340-BEI EINATMEN: Die Person an die frische Luft bringen und für ungehinderte Atmung sorgen.
- P405-Unter Verschluss aufbewahren.
- P501-Inhalt/Behälter zuführen.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'