Learn More
IRX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560261
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NKVTWGARSK DQEDGALFGS DTEGDPEKAE DDEEIDLESI DIDKIDEHDG DQSNEDDEDK AEAPHAPAAP SALARDQGSP LAAADVLKPQ DS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.
The IRXA group includes Irx1, Irx2 and Irx4; the IRXB group is comprises Irx3, Irx5 and Irx6. Irx1 and Irx2 are both widely expressed during development in the lung epithelium and also in the ventricular septum. Irx1 and Irx2 also play a role in digit formation (E11.5-E14.5). The Irx gene family members are each expressed in a distinct pattern during mouse heart development. Specifically, Irx1 and Irx2 are expressed in the ventricular septum and Irx3 is expressed in the ventricular trabeculated myocardium. In addition, Irx4 is expressed in the linear heart tube and the AV canal;.Spezifikation
IRX1 | |
Polyclonal | |
Unconjugated | |
IRX1 | |
Homeodomain protein IRXA1; iroquois homeobox 1; iroquois homeobox protein 1; Iroquois related homeobox 1; Iroquois related homeobox 1 (Drosophila); iroquois-class homeodomain protein IRX-1; Irx1; IRX-5; IRXA1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16371, 79192 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P78414, P81068 | |
IRX1 | |
Recombinant protein corresponding to Human IRX1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Research Use Only