Learn More
ISCU Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563455
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PARLYHKKVV DHYENPRNVG SLDKTSKNVG TGLVGAPACG DVMKLQIQVD EKGKIVDARF K Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Iron-sulfur (Fe-S) clusters are necessary for several mitochondrial enzymes and other subcellular compartment proteins. They contain sulfur and iron, and are created via several steps that include cysteine desulfurases, iron donors, chaperones, and scaffold proteins. This gene encodes the two isomeric forms, ISCU1 and ISCU2, of the Fe-S cluster scaffold protein. Mutations in this gene have been found in patients with myopathy with severe exercise intolerance and myoglobinuria.Spezifikation
ISCU | |
Polyclonal | |
Unconjugated | |
ISCU | |
2310020H20Rik; AA407971; HML; hnifU; iron-sulfur cluster assembly enzyme; iron-sulfur cluster assembly enzyme ISCU, mitochondrial; iron-sulfur cluster scaffold homolog; Iscu; IscU iron-sulfur cluster scaffold homolog; IscU iron-sulfur cluster scaffold homolog (E. coli); ISCU2; ISU2; Nifu; NifU-like N-terminal domain containing; nifU-like N-terminal domain-containing protein; NifU-like protein; NIFUN; RGD1309562 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23479 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H1K1 | |
ISCU | |
Recombinant protein corresponding to Human ISCU | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only