missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISG20L2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA567699
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ISG20L2 Polyclonal antibody specifically detects ISG20L2 in Human samples. It is validated for Western Blot.Spezifikation
ISG20L2 | |
Polyclonal | |
Unconjugated | |
ISG20L2 | |
HSD38; HSD-38; interferon stimulated exonuclease gene 20 like 2; interferon stimulated exonuclease gene 20kDa like 2; interferon stimulated exonuclease gene 20kDa-like 2; interferon-stimulated 20 kDa exonuclease-like 2; ISG20L2 | |
Rabbit | |
affinity chromatography | |
RUO | |
81875 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9H9L3 | |
ISG20L2 | |
Synthesized peptide derived from human ISG20L2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |