missing translation for 'onlineSavingsMsg'
Learn More

ISM2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA558910

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15756781

  • 514.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Immunogen sequence: QVTIKVVEDP QAEVSIDLLA EPSNPPPQDT LSWLPALWSF LWGDYKGEEK DRAPGEKGEE KEEDEDYPSE DIEGEDQE Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 53%.

ISM2 Polyclonal antibody specifically detects ISM2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Spezifikation

Spezifikation

ISM2
Polyclonal
Unconjugated
ISM2
Gm264; ISM2; isthmin 2; isthmin 2 homolog; isthmin 2 homolog (zebrafish); isthmin-2; PSEC0137; TAIL1; thrombospondin and AMOP containing isthmin-like 1; thrombospondin and AMOP domain-containing isthmin-like protein 1; Thrombospondin type-1 domain-containing protein 3; thrombospondin, type I domain-containing 3; thrombospondin, type I, domain containing 3; THSD3
Rabbit
Antigen affinity chromatography
RUO
145501
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q6H9L7
ISM2
Recombinant protein corresponding to Human ISM2
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only