Learn More
ISX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563840
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RGMERNSLGC CEAPKKLSLS FSIEAILKRP ARRSDMDRPE GPGEEGPGEA AASGSGLEKP PKDQPQEGRK Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 57%.
Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the RAXLX homeobox gene family.Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the RAXLX homeobox gene family.Spezifikation
ISX | |
Polyclonal | |
Unconjugated | |
ISX | |
9130012O13Rik; DKFZp781N2395; intestine specific homeobox; intestine-specific homeobox; ISX; MGC138417; pancreas-intestine homeodomain transcription factor; Pix-1; RAX-like homeobox; RAXLX | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91464 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2M1V0 | |
ISX | |
Recombinant protein corresponding to Human ISX | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only