Learn More
ISYNA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552487
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SVLVDFLIGS GLKTMSIVSY NHLGNNDGEN LSAPLQFRSK EVSKSNVVDD MVQSNPVLYT PGEEPDHCVV IKYVPYVGDS KRALDEYTSE LMLGGTNTLV LHNTCEDSLL AAPIMLDLA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.
ISYNA1 Polyclonal antibody specifically detects ISYNA1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ISYNA1 | |
Polyclonal | |
Unconjugated | |
ISYNA1 | |
1300017C10Rik; AU018670; hINO1; Ino1; INOS; Inositol-3-phosphate synthase 1; IPS; IPS 1; IPS-1; Isyna1; MGC133800 protein; MI-1-P synthase; MIP synthase; Myo-inositol 1-phosphate synthase; Myo-inositol 1-phosphate synthase A1; myo-inositol-1-phosphate synthase; testis secretory sperm-binding protein Li 200a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51477 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.11 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NPH2 | |
ISYNA1 | |
Recombinant protein corresponding to Human ISYNA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only