Learn More
ITCH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554329
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LKSDVLLGTA ALDIYETLKS NNMKLEEVVV TLQLGGDKEP TETIGDLSIC LDGLQLESEV VTNGETTCSE NGVSLCLP Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.
ITCH Polyclonal antibody specifically detects ITCH in Human samples. It is validated for Immunohistochemistry.Spezifikation
ITCH | |
Polyclonal | |
Unconjugated | |
ITCH | |
6720481N21Rik; 8030492O04Rik; A130065M08; ADMFD; AIF4; AIP4; atrophin-1 interacting protein 4; Atrophin-1-interacting protein 4; C230047C07Rik; dJ468O1.1; E3 ubiquitin-protein ligase Itchy; E3 ubiquitin-protein ligase Itchy homolog; HECT-type E3 ubiquitin transferase Itchy homolog; ITCH; itch E3 ubiquitin ligase; itchy E3 ubiquitin protein ligase; itchy E3 ubiquitin protein ligase homolog; itchy, E3 ubiquitin protein ligase; NAPP1; NFE2-associated polypeptide 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83737 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96J02 | |
ITCH | |
Recombinant protein corresponding to Human ITCH | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only