Learn More
ITFG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554067
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: VSFKNHSALI TSVVPGDYDG DSQMDVLLTY LPKNYAKSEL GAVIFWGQNQ TLDPNNMTIL NRTFQDEPLI MDFNGDLIPD IFGITNESNQ PQILLGGNLS WHPALTTTSK MRIPHSHAFI DLTEDFTA Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.
ITFG1 Polyclonal antibody specifically detects ITFG1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ITFG1 | |
Polyclonal | |
Unconjugated | |
Itfg1 | |
2310047C21Rik; AI314457; CDA08; CDA08-like protein; D8Wsu49e; hypothetical protein CDA08; integrin alpha FG-GAP repeat containing 1; integrin-alpha FG-GAP repeat-containing protein 1; ITFG1; Linkin; LNKN-1; Protein TIP; T-cell immunomodulatory protein; TIP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81533 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TB96 | |
Itfg1 | |
Recombinant protein corresponding to Human ITFG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only