Learn More
ITFG2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558485
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KVLDASGHHE TLIGEEQRPV FKQHIPANTK VMLISDIDGD GCRELVVGYT DRVVRAFRWE ELGEGPEHLT GQLVSLKKWM LEGQVDSLSV TLGPL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
ITFG2 Polyclonal antibody specifically detects ITFG2 in Human samples. It is validated for Immunohistochemistry.Spezifikation
ITFG2 | |
Polyclonal | |
Unconjugated | |
ITFG2 | |
2700050P07Rik; AI646725; integrin alpha FG-GAP repeat containing 2; integrin-alpha FG-GAP repeat-containing protein 2; integrin-alpha FG-GAP repeat-containing protein 2; KICSTOR complex protein ITFG2; ITFG2; KICSTOR complex protein ITFG2; MDS028; RGD1311944 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55846 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969R8 | |
ITFG2 | |
Recombinant protein corresponding to Human ITFG2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only