missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITPKA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14270-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Regulates inositol phosphate metabolism by phosphorylation of second messenger inositol 1,4,5-trisphosphate to Ins(1,3,4,5)P4. The activity of the inositol 1,4,5-trisphosphate 3-kinase is responsible for regulating the levels of a large number of inositol polyphosphates that are important in cellular signaling. Both calcium/calmodulin and protein phosphorylation mechanisms control its activity. It is also a substrate for the cyclic AMP-dependent protein kinase, calcium/calmodulin- dependent protein kinase II, and protein kinase C in vitro.Spezifikation
ITPKA | |
Polyclonal | |
Unconjugated | |
ITPKA | |
InsP 3 kinase A, IP3 3 kinase A, IP3K A, IP3KA, ITPKA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
228550, 3706, 81677 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17105, P23677, Q8R071 | |
ITPKA | |
ITPKA Fusion Protein Ag5588 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |