missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITPKB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA549796
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total ITPKB protein.
ITPKB Polyclonal antibody specifically detects ITPKB in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
ITPKB | |
Polyclonal | |
Unconjugated | |
ITPKB | |
1110033J02Rik; E130307H12Rik; Inositol 1,4,5-trisphosphate 3-kinase B; inositol-trisphosphate 3-kinase B; insP 3-kinase B; IP3 3-kinase B; IP3-3KB; IP3K; IP3K B; IP3KB; IP3K-B; Itpkb; PIG37; proliferation-inducing protein 37 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
320404, 3707, 54260 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P27987, P42335 | |
ITPKB | |
Synthesized peptide derived from Internal of human ITPKB. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |