missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IWS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA567630
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Predicted to react with Rat samples.
Transcription factor which plays a key role in defining the composition of the RNA polymerase II (RNAPII) elongation complex and in modulating the production of mature mRNA transcripts. Acts as an assembly factor to recruit various factors to the RNAPII elongation complex and is recruited to the complex via binding to the transcription elongation factor SUPT6H bound to the C-terminal domain (CTD) of the RNAPII subunit RPB1 (POLR2A). The SUPT6H:IWS1:CTD complex recruits mRNA export factors (ALYREF/THOC4, EXOSC10) as well as histone modifying enzymes (such as SETD2) to ensure proper mRNA splicing, efficient mRNA export and elongation-coupled H3K36 methylation, a signature chromatin mark of active transcription.Spezifikation
IWS1 | |
Polyclonal | |
Unconjugated | |
IWS1 | |
1700069O15Rik; 3000008H23; AL117927; AW214353; interacts with Spt6; IWS1; IWS1 homolog; IWS1 homolog (S. cerevisiae); IWS1, SUPT6H interacting protein; Iws1l; IWS1-like protein; Protein IWS1 homolog; RGD1304762 | |
Rabbit | |
affinity chromatography | |
RUO | |
291705, 55677 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q3SWT4, Q96ST2 | |
IWS1 | |
Synthesized peptide derived from human IWS1. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |