missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14568-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a regulator of the pancreatic beta-cell function. It is highly similar to JIP-1, a mouse protein known to be a regulator of c-Jun amino-terminal kinase (Mapk8). This protein has been shown to prevent MAPK8 mediated activation of transcription factors, and decrease IL-1 beta and MAP kinase kinase 1 (MEKK1) induced apoptosis in pancreatic beta cells. This protein also functions as a DNA-binding transactivator of the glucose transporter GLUT2. RE1-silencing transcription factor (REST) is reported to repress the expression of this gene in insulin-secreting beta cells. This gene is found to be mutated in a type 2 diabetes family, and thus is thought to be a susceptibility gene for type 2 diabetes.Spezifikation
JIP1 | |
Polyclonal | |
Unconjugated | |
MAPK8IP1 | |
IB 1, IB1, Islet brain 1, JIP 1, JIP1, JNK interacting protein 1, MAPK8IP1, PRKM8IP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116457, 19099, 9479 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9R237, Q9UQF2, Q9WVI9 | |
Mapk8ip1 | |
JIP1 Fusion Protein Ag6150 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |