Learn More
KANSL1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560240
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LLAKHVVKHY GQQMKLSMKH QLPKMKTFHE PTTILGNSLP KCTEIKPEVN TLTAENKLWD DAKNGFARCT AAEIQRFAFS ATGLLSHVEE GLDSDA Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.
C2orf67, also termed as chromosome 2 open reading frame 67, encodes a 705 amino-acid protein. It exists a variant, which encodes 987 amino-acid protein. The C2orf56 may express in lymphoid /immune and nervous system. The specific function of C2orf67 is still non-known.Spezifikation
KANSL1L | |
Polyclonal | |
Unconjugated | |
KANSL1L | |
1110028C15Rik; 1190030G24; AI427598; C2orf67; C430010P07Rik; Kansl1l; KAT8 regulatory NSL complex subunit 1 like; KAT8 regulatory NSL complex subunit 1-like; KAT8 regulatory NSL complex subunit 1-like protein; Kiaa4189; male-specific lethal 1 homolog; mKIAA4189; MSL1v2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151050 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A0AUZ9 | |
KANSL1L | |
Recombinant protein corresponding to Human KANSL1L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only