missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Kappa/Lambda MultiVision Antibody Cocktail

Recommended for Immunohistochemistry (Formalin/paraffin), Eprediatrade; Kappa/Lambda, MultiVision Antibody Cocktail is one marker in a comprehensive antibody portfolio.

Marke:  Epredia™ Lab Vision™ MV-2003-R7

Weitere Details : Netto-Gewicht : 0.01200kg

Artikelnummer. 12602657

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

The antibodies to lambda and kappa (L1C1) light chain are reported as useful in the identification of leukemias, plasmacytomas and certain non-Hodgkin lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant. Kappa-lambda double staining contributes to the observation of clonality of the infiltrate. Kappa-Lambda antibodies are provided in optimal concentration in an easy-to-use format for demonstrating clonality of infilerate. For detection of the primary antibodies a ready-to-use MultiVision polymer detection system consisting of anti-mouse/horseradish peroxidase (HRP) + anti-rabbit/alkaline phosphatase (AP) (Mfr. No. TL-012-MHRA) is available. This cocktail is specially formulated from MultiVision polymers that provide increased sensitivity, time-savings and detection simplicity. The MultiVision AP and HRP polymers are innovative, patented technology. The small amino acid polymer subunits minimize conflicts in binding the target protein resulting in more consistent staining and better signal amplification. Ultimately, this gives the user higher sensitivity and antibody efficiency as well as better signal-to-noise ratios. This system is biotin-free, which prevents background staining found with traditional biotin-based methods. MultiVision polymer detection system is a simple and robust one-step polymer detection system for double staining of kappa light chain and lambda light chain in red and blue, respectively.The MultiVision polymer detection system yields high quality double staining in less than 2 hours on formalin-fixed and paraffin-embedded tissue sections. The kit allows observation of two single-stained cells in blue (Lambda light chain) and red (kappa light chain) and/or co-localization marked by a purple/brownish mixed-color. This MultiVision Kappa -Lambda cocktail along with the MultiVision polymer detection system has also proven to work with formalin-fixed and BMA+MMA (1:1) embedded plastic tissue sections.

Host Species: Kappa — Mouse; Lambda — n/a

Clone: Kappa — L1C1; Lambda — N/A

Isotype: Kappa — Mouse IgG1/κ; Lambda — N/A

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Kappa — Not determined; Lambda — Not determined

Immunogen: Kappa — B-lymphoma cells; Lambda — Lambda light chains purified from human

Molecular Weight: Kappa — 25kDa; Lambda — 25kDa

Positive Control: Tonsil

Cellular Localization: Kappa — Cytoplasmic; Lambda — Cytoplasmic

Recommended for:

  • Immunohistochemistry

General References:

1. Immunoenzyme Multiple Staining Methods. CM van der Loos, Handbook no. 45, BIOS Scientific Publishers, Oxford, UK, 1999. ISBN: 1-85996-187-8 or Springer-Verlag, New York, ISBN: 0-387-91594-x.

Spezifikation

Spezifikation

Kappa/Lambda
Monoclonal/Polyclonal
Unconjugated
Kappa -- B lymphoma cells, Lambda -- Lambda light chains purified from human
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin)
Kappa – L1C1, Lambda – N/A
Mouse, Rabbit
7 mL
Cancer and Tumor Biology
Human
IgG1
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote