missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappa Light Chain Monoclonal Antibody (TB28-2), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marke: Affymetrix eBioscience 63-9970-41
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Mouse Monoclonal Antibody
Kappa Light Chain Monoclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Flow CytometrySpezifikation
Kappa Light Chain | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P01602, P01834, P0DOX7 | |
IGK, IGKC, IGKV1-5 | |
Affinity chromatography | |
RUO | |
28299, 3514, 50802 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
TB28-2 | |
Super Bright 600 | |
IGKC | |
D4Mit8; HCAK1; IgK; Igk@; IGKCD; Immunoglobulin K; Kappa LC; Km | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |