Learn More
- Home
- Products
- Pumps
- Pump Parts and Accessories
- Pump Seals
- Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs
Product Code. 10686635
Change view
Click to view available options
Diameter (Metric) Outside:
17 mm
18 mm
19 mm
21 mm
22 mm
24 mm
29 mm
31.7 dia. x 22mmH
36.6 dia. x 23mmH
37 mm
39.7 dia. x 17mmH
39.7 dia. x 19mmH
42.8 dia. x 19mmH
7.5 dia. x 10mmH
8.3 dia. x 10.0mmH
Diameter (Metric) Inside:
11.3 mm
12.5 mm
13.5 mm
14 mm
15 mm
16.3 mm
17.3 mm
17.5 mm
22 mm
29.8 mm
31.4 dia. x 31mmH
33.3 dia. x 22mmH
38.2 dia. x 23mmH
41.4 dia. x 19mmH
44.4 dia. x 19mmH
9.2 dia. x 10mmH
9.9 dia. x 10.0mmH
Height (Metric):
10 mm
12 mm
13 mm
14 mm
15 mm
16 mm
38 mm
41 mm
48 mm
49 mm
51 mm
Unit Size:
Pack of 100
17 product options available for selection
| Product Code. | Diameter (Metric) Outside | Diameter (Metric) Inside | Height (Metric) | Unit Size |
|---|---|---|---|---|
| 10686635 | 17 mm | 11.3 mm | 10 mm | Pack of 100 |
| 10328624 | 18 mm | 12.5 mm | 12 mm | Pack of 100 |
| 10093514 | 19 mm | 13.5 mm | 12 mm | Pack of 100 |
| 10618175 | 19 mm | 14 mm | 12 mm | Pack of 100 |
| 10257024 | 21 mm | 15 mm | 13 mm | Pack of 100 |
| 10628175 | 22 mm | 17.3 mm | 13 mm | Pack of 100 |
| 10451405 | 24 mm | 17.5 mm | 15 mm | Pack of 100 |
| 10587754 | 22 mm | 16.3 mm | 13 mm | Pack of 100 |
| 10105504 | 29 mm | 22 mm | 16 mm | Pack of 100 |
| 10537794 | 37 mm | 29.8 mm | 16 mm | Pack of 100 |
| 10043384 | 39.7 dia. x 17mmH | 31.4 dia. x 31mmH | 38 mm | Pack of 100 |
| 10155694 | 31.7 dia. x 22mmH | 33.3 dia. x 22mmH | 41 mm | Pack of 100 |
| 10470255 | 36.6 dia. x 23mmH | 38.2 dia. x 23mmH | 48 mm | Pack of 100 |
| 10738405 | 39.7 dia. x 19mmH | 41.4 dia. x 19mmH | 49 mm | Pack of 100 |
| 10388564 | 7.5 dia. x 10mmH | 9.2 dia. x 10mmH | 14 mm | Pack of 100 |
| 10758575 | 42.8 dia. x 19mmH | 44.4 dia. x 19mmH | 51 mm | Pack of 100 |
| 10155584 | 8.3 dia. x 10.0mmH | 9.9 dia. x 10.0mmH | 15 mm | Pack of 100 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
Product Code.
10686635
Supplier
Poeppelmann™
Supplier No.
61000080000
Description
- Universal use as a plug or cap
- Acc. with VDA Band 19 and ISO 16232
- Tapered design ideal for accomodating tolerances
Specifications
Specifications
| Color | Yellow |
| Diameter (Metric) Inside | 11.3 mm |
| For Use With (Application) | Used universally as plug or cap, Tapered design is ideal for accommodating tolerances |
| Height (Metric) | 10 mm |
| Quantity | 100 Pack |
| Diameter (Metric) Outside | 17 mm |
| Material | Polyethylene |
| Type | Closing Plug |