missing translation for 'onlineSavingsMsg'
Learn More

Kartogenin, Tocris Bioscience™

Potently induces chondrogenesis in MSCs

Marke:  RD Systems (Bio-Techne) 4513/10

Weitere Details : Netto-Gewicht : 0.00010kg

Menge. 10mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414864

  • 190.08 € / 10 Milligramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Potently induces differentiation of human mesenchymal stem cells into chondrocytes (EC50 = 100 nM). Reduces disease severity in a mouse model of osteoarthritis; displays protective effects against osteoarthritic stimuli in mature chondrocytes in vitro.
Spezifikation

Chemische Identifikatoren

4727-31-5
317.344
kartogenin, 2-1,1'-biphenyl-4-ylcarbamoyl benzoic acid, 2-4-phenylphenyl carbamoyl benzoic acid, unii-q93bbn11cp, n-biphenyl-4-yl-phthalamic acid, q93bbn11cp, 2-1,1'-biphenyl-4-ylamino carbonyl benzoic acid, 2-4-phenylphenyl carbamoyl benzoic acid, 2-n-4-phenylphenyl carbamoyl benzoic acid, 2-1,1'-biphenyl-4-ylcarbamoyl benzoic acid
2-[(4-phenylphenyl)carbamoyl]benzoic acid
C20H15NO3
SLUINPGXGFUMLL-UHFFFAOYSA-N
2826191
C1=CC=C(C=C1)C2=CC=C(C=C2)NC(=O)C3=CC=CC=C3C(=O)O

Spezifikation

Kartogenin
4727-31-5
kartogenin, 2-1,1'-biphenyl-4-ylcarbamoyl benzoic acid, 2-4-phenylphenyl carbamoyl benzoic acid, unii-q93bbn11cp, n-biphenyl-4-yl-phthalamic acid, q93bbn11cp, 2-1,1'-biphenyl-4-ylamino carbonyl benzoic acid, 2-4-phenylphenyl carbamoyl benzoic acid, 2-n-4-phenylphenyl carbamoyl benzoic acid, 2-1,1'-biphenyl-4-ylcarbamoyl benzoic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)NC(=O)C3=CC=CC=C3C(=O)O
317.344
317.34
10mg
C20H15NO3
SLUINPGXGFUMLL-UHFFFAOYSA-N
2-[(4-phenylphenyl)carbamoyl]benzoic acid
2826191
>98%
Videos