missing translation for 'onlineSavingsMsg'
Learn More

KCNF1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA553225

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15721171

  • 514.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Immunogen sequence: VRYYNKQRVL ETAAKHELEL MELNSSSGGE GKTGGSRSDL DNLPPEPAGK EAPSCSSRLK LSHSDTFIPL LTEEKHHRTR LQSCK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

KCNF1 Polyclonal antibody specifically detects KCNF1 in Human samples. It is validated for Immunocytochemistry.
Spezifikation

Spezifikation

KCNF1
Polyclonal
Unconjugated
Kcnf1
Gm1182; IK8; KCNF; Kcnf1; kH1; KV5.1; potassium channel KH1; potassium channel, voltage-gated modifier subfamily F, member 1; potassium voltage-gated channel modifier subfamily F member 1; potassium voltage-gated channel subfamily F member 1; potassium voltage-gated channel, subfamily F, member 1; voltage-gated potassium channel subunit Kv5.1
Rabbit
Antigen affinity chromatography
RUO
3754
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9H3M0
Kcnf1
Recombinant protein corresponding to Human KCNF1
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only