missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNN3 Recombinant Rabbit Monoclonal Antibody (3G7W0)
Beschreibung
Immunogen Sequence: LSKMQNVMYD LITELNDRSE DLEKQIGSLE SKLEHLTASF NSLPLLIADT LRQQQQQLLS AIIEARGVSV AVGTTHTPIS DSPIGVSSTS FPTPYTSSSS C.
Action potentials in vertebrate neurons are followed by an afterhyperpolarization (AHP) that may persist for several seconds and may have profound consequences for the firing pattern of the neuron. Each component of the AHP is kinetically distinct and is mediated by different calcium-activated potassium channels. KCNN3, a member of the KCNN family of potassium channel genes, encodes a protein that is activated before membrane hyperpolarization and is thought to regulate neuronal excitability by contributing to the slow component of synaptic AHP. The encoded protein is an integral membrane protein that forms a voltage-independent calcium-activated channel with three other calmodulin-binding subunits.
Spezifikation
Spezifikation
| Antigen | KCNN3 |
| Anwendungen | ELISA, Western Blot |
| Klassifikation | Recombinant Monoclonal |
| Klon | 3G7W0 |
| Konzentration | 1.09 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol, 0.05% BSA and 0.05% ProClin 300; pH 7.3 |
| Gen | KCNN3 |
| Gen-Zugriffsnummer | P70605, Q9UGI6 |
| Gen-Alias | hSK3; K3; KCa2.3; Kcnn3; potassium calcium-activated channel subfamily N member 3; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 3; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 3; SK3; SKCa 3; SKCa3; small conductance calcium-activated potassium channel 3; small conductance calcium-activated potassium channel protein 3; small-conductance Ca2+-activated K+ channel 3 |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?