missing translation for 'onlineSavingsMsg'
Learn More

KDEL Polyclonal Antibody, Alexa Fluor™ 488, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA1013A488

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15688028

  • 497.00 € / 50 Mikroliter
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

PA1013A488 detects KDEL from human, rat, mouse and hamster. The PA1013A488 immunogen is a synthetic peptide corresponding to residues C T(643) G E E D T S E K D E L(654) of rat GRP94.

KDEL Polyclonal antibody specifically detects KDEL in Hamster, Human, Mouse, Rat samples. It is validated for Immunocytochemistry.
Spezifikation

Spezifikation

KDEL
Polyclonal
Alexa Fluor 488
Kdelr1
78 kDa glucose-regulated protein; 8030486F04Rik; 94 kDa glucose-regulated protein; 94 kDa glucose-regulated protein {ECO:0000250; AL022860; AU019543; AW215843; baffled; Binding-immunoglobulin protein; BiP; D2Wsu141e; D2Wsu17e; ECGP; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; endoplasmic reticulum resident protein 58; Endoplasmic reticulum resident protein 99; Endoplasmin; endoplasmin {ECO:0000250; endothelial cell (HBMEC) glycoprotein; EP58; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; epididymis secretory sperm binding protein Li 89n; ER lumen protein retaining receptor 1; ER lumen protein-retaining receptor 1; ER protein 58; ERD2; Erd2.1; ERp58; ERp99; glucose regulated protein, 78 kDa; glucose-regulated protein; glucose-regulated protein, 78kDa; gp96; gp96 homolog; GRP 78; Grp78; GRP-78; GRP94; GRP-94; HDEL; heat shock 70 kDa protein 5; heat shock 70kD protein 5; heat shock 70kD protein 5 (gl
Rabbit
Antigen affinity chromatography
RUO
100689305, 100750912, 100773331, 10945, 14828, 22027, 25617, 3309, 361577, 362862, 68137, 7184
4° C, store in dark, DO NOT FREEZE!
Liquid
Immunocytochemistry
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
P06761, P08113, P11021, P14625, P20029, P24390, Q569A6, Q66HD0, Q99JH8
Hsp90b1, HSPA5, Kdelr1
Synthetic peptide corresponding to residues C T(643) G E E D T S E K D E L(654) of rat GRP94
50 μL
Primary
Hamster, Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote