missing translation for 'onlineSavingsMsg'
Learn More

KDM5D Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA540120

Weitere Details : Netto-Gewicht : 0.00910kg

Artikelnummer. 15526811

  • 469.00 € / 100 Mikroliter
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: Jurkat and human cerebellar tissue. Predicted reactivity: Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

KDM5D Polyclonal antibody specifically detects KDM5D in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

KDM5D
Polyclonal
Unconjugated
KDM5D
histocompatibility Y antigen; Histone demethylase JARID1D; HY; H-Y; HYA; JARID1D; Jumonji, AT rich interactive domain 1D (RBP2-like); jumonji/ARID domain-containing protein 1D; KDM5D; KIAA0234; lysine (K)-specific demethylase 5D; lysine demethylase 5D; lysine-specific demethylase 5D; Protein SmcY; selected mouse cDNA on Y, human homolog of; SMC homolog, Y chromosome; SMCY; Smcy homolog, Y-linked
Rabbit
Antigen affinity chromatography
RUO
8284
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Q9BY66
KDM5D
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human KDM5D.
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote