Learn More
KIAA0100 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560118
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EHPVDDIDKM KERAAMNNSF IYIKIPQVPL CVSYKGEKNS VDWGDLNLVL PCLEYHNNTW TWLDFAMAVK RDSRKALVAQ VIKEKLRLKS ATGSEVRGKL ETKSDLN Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
KIAA0100 Polyclonal antibody specifically detects KIAA0100 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
KIAA0100 | |
Polyclonal | |
Unconjugated | |
KIAA0100 | |
Antigen MLAA-22; BCOX; BCOX1; breast cancer overexpressed gene 1; breast cancer-overexpressed gene 1 protein; cancer/testis antigen 101; CT101; FMP27; K0100; KIAA0100; protein KIAA0100; U937-associated antigen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9703 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14667 | |
KIAA0100 | |
Recombinant protein corresponding to Human KIAA0100 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only