Learn More
KIAA0701 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558875
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SVNAGANLQN YGETSPDAIS TNSEGAQENH DDLMSVVVFK ITGVNGEIDI RGEDTEICLQ VNQVTPDQLG NISLRHYLCN RPVGSDQKAV IHS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 78%.
KIAA0701 Polyclonal antibody specifically detects KIAA0701 in Human samples. It is validated for Immunohistochemistry.Spezifikation
KIAA0701 | |
Polyclonal | |
Unconjugated | |
Uhrf1bp1l | |
2010319N22Rik; 4930506D01Rik; AA589495; C80672; E030041M21Rik; Kiaa0701; mKIAA0701; SHIP164; syntaxin 6Habc-interacting protein of 164 kDa; Syntaxin-6 Habc-interacting protein of 164 kDa; UHRF1 (ICBP90) binding protein 1-like; UHRF1 binding protein 1 like; UHRF1 binding protein 1-like; UHRF1-binding protein 1-like; UHRF1BP1L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23074 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A0JNW5 | |
Uhrf1bp1l | |
Recombinant protein corresponding to Human KIAA0701 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only