missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1033 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 51101-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a component of the WASH complex, which functions in the intracellular transport of endosomes. Mutations in this gene have been detected in individuals with autosomal recessive cognitive disability. Alternative splicing results in multiple transcript variants.Spezifikation
KIAA1033 | |
Polyclonal | |
Unconjugated | |
WASHC4 | |
KIAA1033, SWIP, WASH complex subunit 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23325 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q2M389 | |
WASHC4 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |