Learn More
KIAA1033 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561299
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SVHTHLKLDD RNPFKVGMKD LALFFSLNPI RFFNRFIDIR AYVTHYLDKT FYNLTTVALH DWATYSEMRN LATQRYGLVM TEAHLPSQTL EQG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
KIAA1033 Polyclonal antibody specifically detects KIAA1033 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
KIAA1033 | |
Polyclonal | |
Unconjugated | |
WASHC4 | |
A230046K03Rik; AA589518; Kiaa1033; mKIAA1033; MRT43; RIKEN cDNA A230046K03 gene; strumpellin and WASH-interacting protein; SWIP; WASH complex subunit 4; WASH complex subunit 7; WASH complex subunit SWIP; WASHC4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23325 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2M389 | |
WASHC4 | |
Recombinant protein corresponding to Human KIAA1033 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only