Learn More
KIAA1191 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561666
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SGSGDKDRNL SDKWSLFGPR SLQKYDSGSF ATQAYRGAQK PSPLELIRAQ ANRMAEDPAA LKPPKMDIPV MEGKKQPPRA HNLKPRDLNV LTPTGF Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 75%.
KIAA1191 Polyclonal antibody specifically detects KIAA1191 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
KIAA1191 | |
Polyclonal | |
Unconjugated | |
KIAA1191 | |
4833439L19Rik; 4930558H15Rik; brain-derived rescue factor p60MONOX; C81457; flavin monooxygenase motif-containing protein of 33 kDa; flavine monooxygenase motif-containing protein of 33 kDa; KIAA1191; P33monox; p60MONOX; Putative monooxygenase p33MONOX; RIKEN cDNA 4833439L19 gene | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57179 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96A73 | |
KIAA1191 | |
Recombinant protein corresponding to Human KIAA1191 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only