Learn More
KIAA1551 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553091
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LTKLESSPRK LHKDKRQENK HKTFLPVKGN TEKSNMLEFK LCPDILLKNT NSVEERKDVK PHPRKEQAPL QVSGIKSTKE DWLKFVATKK RTQKDSQERD NVNSRLSKRS FSADGFEMLQ NPVKDSKEMF QTY Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 59%.
KIAA1551 Polyclonal antibody specifically detects KIAA1551 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
KIAA1551 | |
Polyclonal | |
Unconjugated | |
KIAA1551 | |
C12orf35; KIAA1551; RESF1; Retroelement silencing factor 1; uncharacterized protein C12orf35; uncharacterized protein KIAA1551; UTA2-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55196 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCM1 | |
KIAA1551 | |
Recombinant protein corresponding to Human KIAA1551 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only