Learn More
KIAA1586 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557630
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NKTTRQASLR KKIREHDVSK AHGKIQDLLK ESTNDSICNL VHKQNNKNID ATVKVFNTVY SLVKHNRPLS DIEGARELQE KNGEVNCLNT RYSATRI Highest antigen sequence identity to the following orthologs: Mouse - 24%, Rat - 24%.
E3 SUMO-protein ligase; facilitates UBE2I/UBC9-mediated SUMO2 modification of target proteins (PubMed:26524493).Spezifikation
KIAA1586 | |
Polyclonal | |
Unconjugated | |
KIAA1586 | |
E3 SUMO-protein ligase KIAA1586; E3 SUMO-protein transferase KIAA1586; KIAA1586 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57691 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCI6 | |
KIAA1586 | |
Recombinant protein corresponding to Human KIAA1586 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only