Learn More
KIAA1755 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562307
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LDPFLADLHQ ASSLLQASIE EFEKADPPGG MQEATRCLSK SKELMEAVLR DPGLLGLQRE GGATLARLQH DASRLDFSPD V Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.
KIAA1755 Polyclonal antibody specifically detects KIAA1755 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
KIAA1755 | |
Polyclonal | |
Unconjugated | |
KIAA1755 | |
KIAA1755; RP5-1054A22.3; uncharacterized protein KIAA1755 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85449 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JYT7 | |
KIAA1755 | |
Recombinant protein corresponding to Human KIAA1755 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only