Learn More
KIAA2026 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551684
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: APGGTTIHTA SAPSNVTSLV GSQFSEPCIQ QKIVINTSTP LAPGTQIMIN GTRFIVPPQG LGAGSHVLLI STNPKYGAPL VLNSGQGIQS TPIDNSAQKI TLASNNSLSG QPLQHPLRSP TK Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.
KIAA2026 Polyclonal antibody specifically detects KIAA2026 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
KIAA2026 | |
Polyclonal | |
Unconjugated | |
KIAA2026 | |
KIAA2026; uncharacterized protein KIAA2026 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
158358 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5HYC2 | |
KIAA2026 | |
Recombinant protein corresponding to Human KIAA2026 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only