Learn More
KIF21B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555563
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: TMKGSTSHDD FKFKSEPKLS AQMKAVSAEC LGPPLDISTK NITKSLASLV EIKEDGVGFS VRDPYYRDRV SRTVSLP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
KIF21B Polyclonal antibody specifically detects KIF21B in Human samples. It is validated for Immunohistochemistry.Spezifikation
KIF21B | |
Polyclonal | |
Unconjugated | |
KIF21B | |
2610511N21Rik; DKFZP434J212; KIAA0449; KIF21B; Kif6; kinesin family member 21B; kinesin-like protein KIF21B; Kinesin-like protein KIF6; LOW QUALITY PROTEIN: kinesin-like protein KIF21B; mKIAA0449; N-5 kinesin; zmp:0000000534 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23046 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75037 | |
KIF21B | |
Recombinant protein corresponding to Human KIF21B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only