Learn More
Thermo Scientific™ KingFisher™ Pure Plasmid Kit
Achieve rapid and reproducible purification of high-quality plasmid DNA from cultured bacteria.
N.V.
Spezifikation
Beschreibung | KingFisher Pure Plasmid Kit, 384 samples |
---|---|
Probentyp | Bacterial Culture |
Geeignete nachgelagerte Analyse | PCR, Real-time PCR, Enzymatic Reactions |
Elutionsvolumen (metrisch) | 100µL |
Beschreibung
Thermo Scientific™ KingFisher ™Pure Plasmid Kit is designed for rapid automated purification of plasmid DNA from cultured bacteria using KingFisher instruments. Plasmid DNA can be purified from 1-5mL of bacterial culture. The excellent performance of KingFisher Plasmid DNA Kit produces plasmid DNA that is free of inhibitors and ready for use in many downstream applications. Easy-to-use KingFisher system and straightforward Thermo Scientific™ BindIt™ protocols facilitate safe sample handling and the maximized walk-away time ensures efficiency.
- Excellent yield, purity and reproducibility
- Flexible KingFisher system
- Walk-away solution for any throughput
- Compatible with most common downstream analysis and application
Spezifikation
KingFisher Pure Plasmid Kit, 384 samples | |
Bacterial Culture | |
PCR, Real-time PCR, Enzymatic Reactions | |
100µL |