Learn More
Thermo Scientific™ KingFisher™ Pure Plasmid Kit
Achieve rapid and reproducible purification of high-quality plasmid DNA from cultured bacteria.
N.V.
Spezifikation
Beschreibung KingFisher Pure Plasmid Kit, 384 samples
Probentyp Bacterial Culture
Geeignete nachgelagerte Analyse PCR, Real-time PCR, Enzymatic Reactions
Elutionsvolumen (metrisch) 100µL
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
Thermo Scientific™ KingFisher ™Pure Plasmid Kit is designed for rapid automated purification of plasmid DNA from cultured bacteria using KingFisher instruments. Plasmid DNA can be purified from 1-5mL of bacterial culture. The excellent performance of KingFisher Plasmid DNA Kit produces plasmid DNA that is free of inhibitors and ready for use in many downstream applications. Easy-to-use KingFisher system and straightforward Thermo Scientific™ BindIt™ protocols facilitate safe sample handling and the maximized walk-away time ensures efficiency.
  • Excellent yield, purity and reproducibility
  • Flexible KingFisher system
  • Walk-away solution for any throughput
  • Compatible with most common downstream analysis and application
Spezifikation
Spezifikation
KingFisher Pure Plasmid Kit, 384 samples
Bacterial Culture
PCR, Real-time PCR, Enzymatic Reactions
100µL