Learn More
Thermo Scientific™ KingFisher™ Pure RNA Plant Kit
Achieve rapid and reproducible purification of high-quality RNA from various plant materials.
N.V.
Spezifikation
Probentyp | Fresh or Frozen Plant Tissues |
---|---|
Geeignete nachgelagerte Analyse | PCR, Real-time PCR, Enzymatic Reactions |
Elutionsvolumen (metrisch) | 100µL |
Beschreibung
Thermo Scientific™ KingFisher™ Pure RNA Plant Kit is designed for rapid automated purification of total RNA from plant samples using KingFisher instruments. Total RNA can be purified from up to 50mg of fresh plant tissue. The excellent performance of KingFisher Pure RNA Plant Kit produces high-integrity RNA that is free of inhibitors and ready for use in many downstream applications. Easy-to-use KingFisher system and straightforward Thermo Scientific™ BindIt™ protocols facilitate safe sample handling and the maximized walk-away time ensures efficiency.
- Excellent yield, purity and reproducibility
- Flexible KingFisher system
- Walk-away solution for any throughput
- Compatible with most common downstream analyses and applications
Spezifikation
Fresh or Frozen Plant Tissues | |
PCR, Real-time PCR, Enzymatic Reactions | |
100µL |