missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KISS1R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA549719
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous level of total KISS1R / GPR54 protein.
KISS1R Polyclonal antibody specifically detects KISS1R in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KISS1R | |
Polyclonal | |
Unconjugated | |
KISS1R | |
AXOR12; CPPB1; G protein-coupled receptor 54; Gpr54; G-protein coupled receptor 54; G-protein coupled receptor OT7T175; HH8; HOT7T175; Hypogonadotropin-1; kiSS-; KiSS-1; KISS1 receptor; kiSS-1 receptor; KISS1R; KiSS-1R; Kisspeptins receptor; metastin receptor; mOT7T175; orphan G protein-coupled receptor 54; orphan G protein-coupled receptor GPR54; rOT7T175 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114229, 78976, 84634 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q91V45, Q924U1, Q969F8 | |
KISS1R | |
Synthetic peptide corresponding to a region corresponding the the C terminal region of Human KISS1 receptor. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |