missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550232
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total KLF15 protein.
KLF15 Polyclonal antibody specifically detects KLF15 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
KLF15 | |
Polyclonal | |
Unconjugated | |
KLF15 | |
1810013I09Rik; AV048136; AW494632; Cardiovascular Krueppel-like factor; CKLF; DKFZp779M1320; Kidney-enriched krueppel-like factor; kidney-enriched Kruppel-like factor; KKLF; KKLF protein; Klf15; KLF15; transcription factor; Krueppel-like factor 15; Kruppel like factor 15; Kruppel-like factor 15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28999 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9UIH9 | |
KLF15 | |
Fusion protein corresponding to residues near the N terminal of human Kruppel-like factor 15 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |