Learn More
KLHDC5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554292
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: CKPQFHLLGS PPQAPGDVSL KQRLREARMT GTPVLVALGD FLGGPLAPHP YQGEPPSMLR YEEMTERWFP LANNLPPDLV NVRGYG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
KLHDC5 Polyclonal antibody specifically detects KLHDC5 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
KLHDC5 | |
Polyclonal | |
Unconjugated | |
KLHL42 | |
A330079N23Rik; AI450355; C230080I20Rik; Ctb9; cullin-3-binding protein 9; kelch domain containing 5; kelch domain-containing protein 5; kelch like family member 42; kelch-like 42; kelch-like family member 42; kelch-like family, member 42; kelch-like protein 42; KIAA1340; Klhdc5; KLHL42; mKIAA1340 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57542 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2K6 | |
KLHL42 | |
Recombinant protein corresponding to Human KLHDC5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only