missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KPNA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10819-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The import of proteins into the nucleus is a process that involves at least 2 steps. The first is an energy-independent docking of the protein to the nuclear envelope and the second is an energy-dependent translocation through the nuclear pore complex. Imported proteins require a nuclear localization sequence which generally consists of a short region of basic amino acids or 2 such regions spaced about 10 amino acids apart. Proteins involved in the first step of nuclear import have been identified in different systems. These include the Xenopus protein importin and its yeast homolog, SRP1, which bind to the NLS. KPNA2 protein interacts with the NLSs of DNA helicase Q1 and SV40 T antigen and may be involved in the nuclear transport of proteins. KPNA2 also may play a role in VJ recombination.Spezifikation
KPNA2 | |
Polyclonal | |
Unconjugated | |
KPNA2 | |
Importin subunit alpha 2, IPOA1, Karyopherin subunit alpha 2, KPNA2, QIP2, RAG cohort protein 1, RCH1, SRP1, SRP1 alpha, SRP1alpha | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16647, 3838, 85245 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P52292, P52293 | |
KPNA2 | |
KPNA2 Fusion Protein Ag1153 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |