missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 23 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550198
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total KRT23 protein.
Cytokeratin 23 Polyclonal antibody specifically detects Cytokeratin 23 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Cytokeratin 23 | |
Polyclonal | |
Unconjugated | |
KRT23 | |
C HAIK1; CK23; CK-23; cytokeratin 23; cytokeratin-23; DKFZP434G032; HAIK1; histone deacetylase inducible keratin 23; hyperacetylation-inducible type I keratin; K23; Ka23; keratin 23; keratin 23 (histone deacetylase inducible); keratin 23, type I; keratin complex 1, acidic, gene 23; keratin, type I cytoskeletal 23; Keratin-23; Krt1-23; KRT23; MGC26158; type I intermediate filament cytokeratin; type I keratin KA23 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25984 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9C075 | |
KRT23 | |
Fusion protein corresponding to residues near the N terminal of human keratin 23 (histone deacetylase inducible) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |