missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT77 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA567833
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. KRT77 encodes an epithelial keratin that is expressed in the skin and eccrine sweat glands. The type II keratins are clustered in a region of chromosome 12q13.Spezifikation
KRT77 | |
Polyclonal | |
Unconjugated | |
KRT77 | |
KRT1B | |
Rabbit | |
Affinity chromatography | |
RUO | |
374454 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q7Z794 | |
KRT77 | |
Synthesized peptide derived from human KRT77. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |