missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRTAP20-3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562815
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DCKYSYTSGF GAFRILDCGY RCGCGG Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 50%.
KRTAP20-3 Polyclonal antibody specifically detects KRTAP20-3 in Human samples. It is validated for Immunohistochemistry.Spezifikation
KRTAP20-3 | |
Polyclonal | |
Unconjugated | |
KRTAP20-3 | |
KAP19D; KAP20.3; keratin associated protein 19 pseudogene 4; keratin associated protein 20-3; keratin-associated protein 20-3; KRTAP19P4; KRTAP20-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
337985 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3LI60 | |
KRTAP20-3 | |
Recombinant protein corresponding to Human KRTAP20-3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only