Learn More
KTN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553652
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ELKRLEAMLK ERESDLSSKT QLLQDVQDEN KLFKSQIEQL KQQNYQQASS FPPHEELLKV ISEREKEISG LWNELDSLKD AVEHQRKKNN ERQQQVEAVE LEAKE Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 57%.
KTN1 Polyclonal antibody specifically detects KTN1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
KTN1 | |
Polyclonal | |
Unconjugated | |
KTN1 | |
CG1; CG-1 antigen; KIAA0004; Kinectin; kinectin 1; kinectin 1 (kinesin receptor); kinesin receptor; KNT; ktn1; MU-RMS-40.19 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3895 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86UP2 | |
KTN1 | |
Recombinant protein corresponding to Human KTN1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only