Learn More
KXD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559602
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ARQHPEAFSH IPEASFLEEE DEDPIPPSTT TTIATSEQST GSCDTSPDTV SPSLSPGFED LSHVQAGSPA INGRSQTDDE EMTGE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 85%.
KXD1 Polyclonal antibody specifically detects KXD1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KXD1 | |
Polyclonal | |
Unconjugated | |
KXD1 | |
BORCS4; C19orf50; KXD1; KXDL; KxDL motif containing 1; kxDL motif-containing protein 1; MST096; MSTP096; UPF0459 protein C19orf50 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79036 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BQD3 | |
KXD1 | |
Recombinant protein corresponding to Human KXD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only