Learn More
KY Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557686
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: AQGTLSDQQA NPSSLLQRGG GFQGVGNGVR RWQKLEGNDF HENLVEKQHP QQPQVITSYN SQGTQLTVEV HPRDAMPQ Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 69%.
KY Polyclonal antibody specifically detects KY in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
KY | |
Polyclonal | |
Unconjugated | |
KY | |
CD1; D9Mgc44e; ky; Kyphoscoliosis peptidase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
339855 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NBH2 | |
KY | |
Recombinant protein corresponding to Human KY | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only