missing translation for 'onlineSavingsMsg'
Learn More

Avantor™ L(+)-Histidine, USP, Multi-Compendial, GMP, J.T.Baker™

Product Code. 18887059
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18887059 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18887059 Supplier Avantor™ Supplier No. 208006
 Restricted Item
Verfügbar ab: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L(+)-Histidine
Melting Point 272°C to 273°C
CAS 71-00-1
pH 7 to 8.5
Color White
Molecular Formula C6H9N3O2
Packaging Poly bottle, tamper-evident cap
Synonym L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773, L-Histidine
Molecular Weight (g/mol) 155.16
Quantity 1 kg
Percent Purity 98.5 to 101.5%
Grade Multi-Compendial/USP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.